Would you like to log in again?
Mall Probe, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63324-20
102094-608EA 5.26 USD InStock
102094-608
Mall Probe, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Mall Probe: 6” long, Chrome plated with one blunt bent end and hexagonal shape handle
Probe with Eye: Stainless steel, 5” long, 2 mm diameter. Flat one end with eye.
Huber Probe: Chrome Plated. 6” long, one blunt bent end, and one chisel finish on the other end
Inoculating Loop/Needle: Nichrome wire. An 8" aluminum handle with a 3" nichrome wire, 25 gauge. Loop outside diameter is 4 mm. Loop inside diameter is 2.5 mm. Available with a loop or a needle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty