Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
NUNC™ MaxiSorp™ 96 Well Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63494-BK 63494-BK 63494-WH 63494-CL
102094-772CS 990.98 USD InStock
102094-772 102094-770 102094-769 102094-768
NUNC™ MaxiSorp™ 96 Well Plates, Electron Microscopy Sciences
Microplates Microarray Plates
NUNC MaxiSorp plates have been the certified* ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing.
  • Hydrophilic MaxiSorp™ surface binds antibodies, protein and biomolecules
  • Flat bottom, solid polystyrene plates
  • Excellent for chemiluminescent, fluorescent and luminescent arrays
  • Standard plate footprint is compatible with exiting readers
* Certified for absorption of IgG; CV of less than 5% between wells. All results are ±10% from the mean and background of all well ±0.005 absorbance units from the mean.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty