Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
TruBond™ Adhesion Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63701-W10 63701-B10 63700-W1 63701-Y10 63702-G1 63703-G1 63703-Y10 63702-G10 63701-G10 63700-Y10 63701-G1 63702-B1 63702-Y10 63700-G1 63703-G10 63700-B1 63703-Y1 63700-W10 63702-Y1 63701-B1 63703-W10 63702-W10 63702-B10 63703-B1 63700-B10 63700-G10 63701-Y1 63701-W1 63702-W1 63703-B10 63700-Y1
102094-796BX 78.05 USD InStock
102094-796 102094-850 102094-808 102094-828 102094-806 102094-804 102094-826 102094-848 102094-802 102094-824 102094-846 102094-822 102094-800 102094-844 102094-842 102094-820 102094-840 102094-818 102094-816 102094-838 102094-814 102094-858 102094-836 102094-834 102094-856 102094-812 102094-832 102094-810 102094-854 102094-798 102094-830
TruBond™ Adhesion Slides, Electron Microscopy Sciences
Slides Adhesive Slides
TruBond™ adhesive slides have proven performance under high temperature and pH conditions. These adhesive slides work well with automatic IHC applications. The quality of these slides will exceed your expectations.
Ordering information: All styles available in white, blue, green, and yellow. Pink is available in TruBond™ 360 and 200 only.
Packaging: TruBond™ is packaged 100 slides per box, 10 boxes to a case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty