Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
63841-10 63826-10 63832-10 63825-10 63843-10 63840-10 63823-05 63825-05 63822-05 63827-05 63842-10 63843-05 63820-10 63829-10 63844-05 63844-10 63840-05 63824-05 63831-10 63828-10 63833-10 63823-10 63841-05 63820-05 63821-05 63827-10 63830-10 63826-05 63821-10 63842-05 63822-10 63824-10
102094-872PK 106.9 USD InStock
102094-872 102094-894 102094-892 102094-870 102094-890 102094-908 102094-906 102094-904 102094-902 102094-868 102094-900 102094-922 102094-888 102094-866 102094-920 102094-886 102094-864 102094-862 102094-884 102094-882 102094-860 102094-880 102094-918 102094-916 102094-914 102094-912 102094-878 102094-910 102094-876 102094-898 102094-874 102094-896
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Slides Microscope Slides
Made from Corning® 7740 borosilicate, low expansion, type I glass. Micro glass slides are precision rectangular glass capillary tubes for use in microscopic viewing of liquid samples.

  • packaged in plastic vials
  • Internal diameter tolerance is ±10%
  • Wall tolerance is ±20%
  • Withstand heat up to 500 °C

Capillary action instantly collects the sample and the thin glass walls minimize evaporation. The design of these cells eliminates the needs for cover glass and the hanging drop method. Samples can be retained for future reference by sealing or cementing open ends.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty