Would you like to log in again?
BOP Starter Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63920-50 63920-60 63920-01
102094-928EA 679.79 USD InStock
102094-928 102094-926 102094-924
BOP Starter Kit, Electron Microscopy Sciences
Boxes Cryoboxes
Introducing the world’s first single specimen container for multiple endoscopic biopsies.
Simply stated, a BOP kit is a secure plate of specialized, easy to open and reseal biopsy specimen containers in an anatomical map format.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty