Would you like to log in again?
RDO Rapid Decalcifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
64144-40 64143-40 64143-06 64143-01 64144-06 64144-44 64143-44 64144-01
102094-950EA 164.89 USD InStock
102094-950 102094-938 102094-948 102094-946 102094-944 102094-942 102094-952 102094-940
RDO Rapid Decalcifier, Electron Microscopy Sciences
Stains Microscopy and staining reagents
RDO and RDO Gold were developed with the basic premise of making the work routine quicker and easier without sacraficing the quality and integrity of samples. These are remarkably effective multipurpose decacifiers that can be tailored to suit specific lab routine.
  • Safe, non toxic and non-corrosive
  • Dilutable and translucent
  • Microwavable and biodegradable
  • Great cellular differentiation
Both RDO and RDO gold provides crisp nuclear staining and definition, give distinguishable chromatin, show great surface decalcification, safe enough to be disposed down to lab sink to drain.
RDO Gold is designated for biopsies which contain minimal amounts of calcium. It requires no filtration and gives stellar morphological representation of bone biopsies. There is no folding of tissues with RDO Gold and it is clear and non-precipitate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty