Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64707-01 64705-01 64705-05 64707-05 64706-05 64706-01
102095-520PK 422.08 USD InStock
102095-520 102095-530 102095-526 102095-528 102095-522 102095-524
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
Microplates Cell Culture Plates
Tissue culture plates are designed and treated for a wide range of applications. Plates are made from optically clear, medical grade polystyrene with flat bottoms of uniform thickness, allowing for distortion-free observation and microscopic examination. Alphanumeric coordinates are placed on the same focal plane as cell growth-for easy referencing of well position. These plates are also for use with our NetWells and our MeshWell Plates.
  • Maintenance of stock cultures
  • Cell cloning
  • Hybridoma fusions
  • Scale up culture
  • Clone expansion
  • 3H-Thymidine
  • 51Cr Release assay
  • MTT assays
  • In vitro toxicology
  • Drug binding studies
  • Virus isolation
  • Viral plaque assay
  • Transformation
  • Carcinogenicity testing
  • Mutagenicity testing
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty