Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Dounce Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64790-01 64790-40 64790-15 64790-07
102095-540PK 575.74 USD InStock
102095-540 102095-542 102095-538 102095-544
Dounce Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
This grinder designed to retain a high percentage of cell nuclei and mitochondria in soft tissues from cell cultures. When use, the pestle ball is encircled in liquid, which is avoid heat buildup by reducing friction.
  • Ideal for soft tissue
  • Provide with loose and tight pestle
  • Wheaton 33 low extractable borosilicate glass
  • Autoclavable
This grinder is ideal for enzyme studies. The initial grinding is performed using the “loose” pestle. The grinding process is completed using the “tight” pestle.. This grinder is used by moving the pestle up and down, and work best with cell suspensions or very soft tissues. The mortar has a large reservoir and pouring lip and is supplied with “loose” and “tight” pestles. Tight Pestle “A” Specification: 0.0010 – 0.0030” (0.025 – 0.076 mm) Loose Pestle “B” Specification: 0.0035 – 0.0055” (0.089 – 0.14 mm)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty