Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64792-74 64792-02 64792-82 64792-72 64792-80 64792-05 64792-15 64792-73 64792-71 64792-84 64792-30 64792-81 64792-75 64792-83 64792-85 64792-10 64792-70 64792-55
102095-558PK 480.9 USD InStock
102095-558 102095-566 102095-576 102095-554 102095-568 102095-578 102095-556 102095-584 102095-562 102095-572 102095-586 102095-564 102095-552 102095-574 102095-580 102095-560 102095-582 102095-570
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. They are designed for use in preparation of tissue homogenates and other similar applications.
  • Ideal for tissue homogenates
  • Use with power homogenization
  • Parts interchangeable
  • Teflon® pestle
  • Autoclavable
The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3mm) diameter. The grinding chamber clearance is 0.004" to 0.006’ (0.1 to 0.15mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer. However, we strongly recommend the use of safety-coated tissue grinder (64793-series) for added safety. This unit is manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type 1 and ASTM E-438 Type 1, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty