Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Potter-Elvehjem Safe-Grind Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64793-72 64793-73 64793-10 64793-70 64793-30 64793-02 64793-55 64793-05 64793-71 64793-74 64793-15 64793-75
102095-594PK 479.48 USD InStock
102095-594 102095-596 102095-606 102095-590 102095-592 102095-608 102095-602 102095-604 102095-588 102095-610 102095-598 102095-600
Potter-Elvehjem Safe-Grind Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
Wheaton Safe-grind plastic coated tissue grinders afford an added measure of safety over uncoated glass tissue grinders.
  • Exterior plastic coated glass mortar provide greater safety
  • All features other features the same as 64792-series
The heavy plastic coating protects the mortar from becoming scratched or checked, and assures you of sure grip. The transparent coating allows an unobstructed view of the homogenate during grinding. The grinders are fully autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty