Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64794-70 64794-01 64794-80
102095-614PK 219.94 USD InStock
102095-614 102095-616 102095-612
Tissue Grinder, 0.1 ml, Electron Microscopy Sciences
Homogenizers Tissue Grinders
Similar to the all glass Potter-Elvehjem style, but micro size. Ideal for extremely precise work.
  • Made from Wheaton 33 low extractable borosilicate glass
  • Micro size for extremely precise work
  • Reservoir and pouring lip are incorporated into the design
  • Autoclavable
This unit is designed for delicate hand operation. Clearance between pestle and tube is 0.004” to 0.006” (0.1 – 015 mm).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty