Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64795-80 64795-30 64795-02 64795-05 64795-84 64795-83 64795-15 64795-82 64795-85 64795-55 64795-10 64795-81
102095-630PK 246.41 USD InStock
102095-630 102095-640 102095-618 102095-628 102095-636 102095-624 102095-638 102095-626 102095-632 102095-620 102095-634 102095-622
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty