Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Tapered Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64796-01 64796-15 64796-03 64796-05
102095-646EA 233.05 USD InStock
102095-646 102095-648 102095-642 102095-644
Tapered Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
Grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require.
  • Homogenizes connective tissue including heart, muscle and lung
  • Made from Wheaton 33 low extractable borosilicate glass
  • Interchangeable components takes the hassle out of matching parts
  • Autoclavable
The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step.The components are interchangeable. Therefore no need keep parts matched during cleaning. The ground glass surface on both the mortar and pestle are capable of homogenizing connective tissue such as heart, muscle and lung, as well as skin and plant tissue.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty