Would you like to log in again?
Tapered Tissue Grinder Stainless Steel Rod Pestle, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
64797-05 64797-71 64797-03 64797-72 64797-80 64797-83 64797-70 64797-15 64797-81 64797-82 64797-01 64797-73
102095-672EA 273.63 USD InStock
102095-672 102095-682 102095-674 102095-684 102095-670 102095-680 102095-668 102095-676 102095-686 102095-678 102095-688 102095-666
Tapered Tissue Grinder Stainless Steel Rod Pestle, Electron Microscopy Sciences
Homogenizers Tissue Grinders
This unit is the same as the 64796-series, but with a Teflon® pestle for soft tissues.
  • Teflon pestle with stainless steel rod firs power homogenizer
  • Homogenizes soft tissue such as brain
  • Mortar made from Wheaton 333 low extractable borosilicate glass
  • Interchangeable components
  • Autoclavable
The stainless steel rod is 0.25” (6.3 mm) diameter and is suit for use with Wheaton Overhead Stirrer.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty