Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nunc® Nunclon™ Multi-Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64810-17 64811-10 64810-16 64810-05 64811-05 64810-18 64810-19 64810-15
100500-496CS 409.06 USD InStock
100500-496 100500-478 102095-724 102095-726 102095-720 100500-482 102095-722 100500-494
Nunc® Nunclon™ Multi-Dishes, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black for fluorometric detection. External dimension: 128 x 86 mm.
  • White or black upper structure with Polymer bottom combines the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications
  • Optical clarity for viewing well contents
  • Footprint compatible with standard equipment and automated systems
  • Flat bottom well geometry for plate reader access
  • Cell culture, poly-D-Lysine or Collagen I surfaces for cell-based array
  • Non-treated plates are optimized for scintillation counting
  • Working ranges: 50 –200 µl/well
Polystyrene lids have external dimensions of 128 x 86 mm.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty