Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nunc™ 96 Well Optical Bottom Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64816-15 64815-15 64815-10 64815-20 64815-05
102095-740CS 4022.96 USD InStock
102095-740 100490-668 100500-506 100490-658 100500-508
Nunc™ 96 Well Optical Bottom Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black for fluorometric detection. Sterile. External dimensions 128 x 86 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty