Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eyepiece Graticules, Scales with Crosslines, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68012-19 68013-23 68013-25 68013-24.5 68013-21 68013-20.4 68012-21 67980-19 68013-27 67980-50 68013-19 68012-16 68013-26 67980-21 68013-16 67980-16
103305-064EA 246.12 USD InStock
103305-064 102096-042 103305-066 103305-060 102096-046 103305-062 102096-044 103305-056 103305-058 100499-156 100499-158 100499-152 100499-154 102096-048 100499-160 100499-162
Eyepiece Graticules, Scales with Crosslines, Electron Microscopy Sciences
Graticules Reticles and Grids
Useful when interested in measurement in different axis.
  • Used as a horizontal and a vertical scale
The microscope graticle is used as a horizontal and a vertical scale and is especially useful when measuring two axis.
NE7 : Horizontal micrometer scale 10 mm (0.393 inches) long with crosslines at 0.1 mm (0.003 inches) spacing.
NE77 : Horizontal micrometer scale 5 mm (0.196 inches) long in 100 divisions with crosslines.
NE777 : Horizontal micrometer scale12.7 mm (0.5 inches) long with divisions of 0.127 mm (0.005 inches) and crosslines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty