Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Grain Sizing Reticles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68036-27 68007-50 68036-50 68007-26 68036-26 68007-27 68007-23 68007-25 68036-24 68007-21 68036-25 68036-21 68036-23 68007-24
102096-122EA 581.89 USD InStock
102096-122 102096-100 102096-120 103306-240 102096-116 103306-244 102096-104 102096-114 102096-102 103306-242 103306-238 102096-108 102096-118 102096-106
Grain Sizing Reticles, Electron Microscopy Sciences
Graticules Reticles and Grids
Determination of non-metallic inclusion content of steel.
  • Scaled for use with ten objective magnification
Certifications: NG60 meets EN 10247 and NG61 Meets ISO 4976.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty