Would you like to log in again?
Star Test Chart Sector, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68098-09 68098-08
102096-174EA 366.05 USD InStock
102096-174 102096-172
Star Test Chart Sector, Electron Microscopy Sciences
Charts
Star Test Chart Sector are Image type, Vacuum deposited chrome.
R65 Star Test Target's key features are the Siemens Star or Sunburst Pattern
The R66 Star Test Target also has the Siemens Star or Sunburst Pattern. It has the pattern R66, 72 Sectors, and 5° pitch. Outside diameter of image, 25 mm. Unresolved centre core, 0.4 mm. Normal substrate, B270 Glass. Overall size, 50 x 50 mm.
In addition to the pattern R65 and sector pitch 10°, the R65 Star Test Target has thirty-six sectors. Outside diameter of image, 25 mm. Unresolved centre core, 0.2 mm. Normal substrate B270 Glass. Overall size 50 x 50 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty