Would you like to log in again?
NBS 5 Resolution Chart, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68098-11
102096-178EA 393.31 USD InStock
102096-178
NBS 5 Resolution Chart, Electron Microscopy Sciences
Charts
A series of gratings diminishing in size as they go towards the central target. Lines range from 3 lines per mm to 20 lines per mm. Supplied as a positive high contrast chrome image on glass substrate 50 x 50 mm.
  • 3-30 Lines per mm Chrome surface image B270 Glass 50 mm x 50 mm
Also available in low contrast and negative form.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty