Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Grid Resolution Chart, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68098-23 68098-22 68098-21 68098-20 68098-24
102096-184EA 393.32 USD InStock
102096-184 102096-182 102096-188 102096-186 102096-180
Grid Resolution Chart, Electron Microscopy Sciences
Charts
Consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 x 75 mm glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty