Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Gratings-Ronchi Rulings, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68098-26 68098-29 68098-25 68098-32 68098-27 68098-31 68098-30 68098-28
102096-196EA 550.65 USD InStock
102096-196 102096-194 102096-200 102096-198 102096-204 102096-202 102096-192 102096-190
Gratings-Ronchi Rulings, Electron Microscopy Sciences
Graticules Reticles and Grids
The grating covers the entire plate with the exception of R25, R26 and R27 where the grating occupies a 30 mm wide central strip. The glass is B270 with refractive index of 1.52.
  • Paired as light gates
  • Demonstrate interference patterns
  • Produce optical gradients
Metallic grating pattern on glass squares with opaque lines approximately 50% of the pitch. Stock items are on 50×50 mm squares of glass 1.5 mm thick.
Mutually perpendicular pairs produce a screen of square apertures. Vacuum deposited chrome gratings on glass substrate. Typically used for testing of resolution, field distortion and parfocal stability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty