Would you like to log in again?
Synergel™ Agarose Additive, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68500-10
102096-302EA 151.85 USD InStock
102096-302
Synergel™ Agarose Additive, Electron Microscopy Sciences
Electrophoresis Reagents Western Blotting Reagents
Improving Agarose Gel Performance
  • Easy To Use – Mix, boil and pour!
Superior Sieving – Better sieving than agarose alone, 1 gr Synergel = 2 gr agarose
High Resolution – Synergel / Agarose gels exhibit improved spatial separation and tighter banding for DNA fragments of 50-2000bp
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty