Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69910-01 69910-10 69910-05
102096-534PK 58.35 USD InStock
102096-534 102096-532 102096-536
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Cassettes Tissue Capsules
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty