Would you like to log in again?
eCount Colony Counter, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70309-01
102096-602EA 487.42 USD InStock
102096-602
eCount Colony Counter, Electron Microscopy Sciences
Colony Counters
The eCount is a simple to use colony counter for counting cells inmicrobiology applications or tissue culture. This unit combines a multifunctionelectronic counter with a Sharpie® pen for counting and markingto prevent missing or double-counting colonies in a Petri dish.
  • Save - Power off button helps save battery life
  • Verify - Each count verified with a beep or LED flash
  • Accurate - Count up and then count down to verify count accuracy
  • Easy To Read - Display up to 32 counts
  • Memory - Keep up to 32 different counts in memory
  • Secure - Disable counter and use the marker to write notes and totals
Delivery information: Package includes: eCount colony counter, black Sharpie® pen for highly visible marks on both plastic and glass, ong-life button battery, hand tally adapter counts without marking and weighted stand for convenient bench top use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty