Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Oncyte® Nitrocellulose Film Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70335-56N 70335-70A 70335-59A 70335-72N 70335-61N 70335-50N 70335-59N 70335-72A 70335-71A 70335-57N 70335-50A 70335-71N 70335-53N 70335-62N 70335-56A 70335-55N 70335-54N 70335-60N 70335-58N 70335-52N 70335-63A 70335-70N 70335-53A 70335-55A 70335-54A 70335-52A 70335-58A 70335-51A 70335-62A 70335-61A 70335-57A 70335-51N 70335-63N 70335-60A
102096-760PK 356.43 USD InStock
102096-760 102096-718 102096-716 102096-738 102096-732 102096-754 102096-710 102096-698 102096-730 102096-752 102096-696 102096-758 102096-736 102096-714 102096-756 102096-712 102096-734 102096-750 102096-694 102096-728 102096-706 102096-704 102096-726 102096-748 102096-708 102096-720 102096-742 102096-740 102096-724 102096-746 102096-702 102096-744 102096-722 102096-700
Oncyte® Nitrocellulose Film Slides, Electron Microscopy Sciences
Slides Microarray Slides
ONCYTE® is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding
Nitrocellulose film surfaces are manufactured and quality tested to the most exacting standards. To provide an optimal balance between binding capacity and fluorescence sensitivity, two variants of nitrocellulose coatings available:
The ONCYTE® AVID™ nitrocellulose-coated substrates are for applications that require the highest possible protein binding affinity and capacity. Therefore AVID™ slides are characterized by unparalleled protein binding capacity and affinity, making them ideal for colometric and infrared applications that require extremely high levels of binding.
The ONCYTE® NOVA™ nitrocellulose-coated substrates are recommended for applications that require the greatest fluorescent sensitivity but can accept reduced binding capacity. Therefore NOVA™ slides extremely low background fluorescence and an elegant, clean surface make them the most sensitive slides available for fluorescence – dependent arrays.
It is also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. Nitrocellulose-coated (NC) slides for protein microarrays, cell lysate arrays, tissue printing and other applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty