Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
- Silicone isolators are available in silicone thickness 0.5 to 2.5 mm
- Isolators are pre-cut into gaskets or as sheet materials with and without SecureSeal™ clean release adhesive on one or both surfaces
- Isolators remain sealed to smooth surfaces
- Non-adhesive isolators are autoclavable and reusable
- Closed chambers may be formed using flexible RNase free, HybriSlip™ covers
They are available with 3 different gasket types, Silicone (S) / Silicone (S) – press-to-seal silicone gasket only, no adhesive, Silicone (S) / Adhesive (A) – press-to-seal, and Adhesive (A) / Adhesive (A) – press-to-seal adhesive on both sides. We also offer the Sheet Material that can be used to match your own design chambers.
Packaging: Isolators are available without coverslips, and packaged 25 per pack.