Would you like to log in again?
Press-To-Seal Silicone Isolators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70347-10 70339-42 70337-70 70339-70 70345-70 70337-72 70347-72 70336-46 70344-20 70347-05 70336-25 70336-20 70336-01 70345-05 70336-02 70348-74 70336-64 70336-76 70346-05 70348-05 70336-32 70344-38 70346-70 70345-39 70345-20 70339-74 70339-05 70348-44 70344-68 70344-39 70346-74 70339-46 70345-10 70344-10 70347-42 70337-25 70339-72 70336-63 70336-03 70336-72 70336-40 70336-31 70344-05 70336-61 70345-40 70336-44 70339-44 70337-42 70336-33 70339-40 70347-44 70337-74 70339-10 70345-38 70337-40 70336-70 70346-42 70336-05 70336-42 70337-76 70347-70 70346-40 70345-68 70346-72 70344-70 70344-40 70347-20 70339-20 70348-70 70348-20 70339-25 70348-10 70339-76 70348-42 70344-69 70336-62 70337-20 70348-40 70337-05 70337-44 70336-10 70347-74 70347-40 70336-04 70336-34 70348-72 70346-20 70337-46 70345-69 70336-74 70337-10 70346-44 70346-10
100490-860PK 231.07 USD InStock
100490-860 102096-882 102096-880 100502-784 102096-838 102096-836 100490-936 100490-858 100490-932 102096-830 100490-854 102096-874 100490-934 100490-856 102096-872 100490-850 102096-878 102096-834 100490-930 100490-852 102096-876 102096-832 100490-870 102096-772 102096-770 102096-806 102096-848 102096-808 100490-866 102096-842 100490-868 102096-840 102096-846 100490-862 102096-844 100490-864 100490-880 100490-882 102096-860 102096-782 102096-780 100502-048 100502-044 100502-046 100490-918 102096-816 102096-814 102096-858 100490-914 100490-916 102096-818 102096-776 102096-852 100490-876 100490-912 102096-774 102096-850 100490-878 102096-812 102096-856 100490-872 102096-778 102096-810 100490-874 102096-854 102096-790 100502-050 100501-308 102096-870 102096-792 100501-306 102096-828 100490-928 102096-826 100490-848 100490-924 100490-926 102096-820 102096-864 100490-920 100501-304 102096-786 102096-862 100490-846 100490-922 102096-784 102096-824 100490-884 102096-868 102096-866 102096-822 100490-886 102096-788
Press-To-Seal Silicone Isolators, Electron Microscopy Sciences
Slides Chamber Slides
Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
  • Silicone isolators are available in silicone thickness 0.5 to 2.5 mm
  • Isolators are pre-cut into gaskets or as sheet materials with and without SecureSeal™ clean release adhesive on one or both surfaces
  • Isolators remain sealed to smooth surfaces
  • Non-adhesive isolators are autoclavable and reusable
  • Closed chambers may be formed using flexible RNase free, HybriSlip™ covers
They are available with 3 different gasket types, Silicone (S) / Silicone (S) – press-to-seal silicone gasket only, no adhesive, Silicone (S) / Adhesive (A) – press-to-seal, and Adhesive (A) / Adhesive (A) – press-to-seal adhesive on both sides. We also offer the Sheet Material that can be used to match your own design chambers.
Packaging: Isolators are available without coverslips, and packaged 25 per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty