Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Disposable Liquid-Holding Slide Wells, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
70366-12 70366-13
102096-886PK 94.33 USD InStock
102096-886 102096-884
Disposable Liquid-Holding Slide Wells, Electron Microscopy Sciences
Slides Chamber Slides
Disposable liquid-holding wells are conveniently pre-cut

  • Examine specimens with or without cover slips
  • Available in depths of 0.25 and 0.50 mm
  • Multiple applications: examine or count tissue culture cells, yeast, bacteria, and other microorganisms
  • Piggy-back with Microscope Slide-Grids to facilitate counting and sizing
  • Compatible with different styles of microscopes (conventional, inverted, or dissecting)
  • Chemically Inert: made of polyester with an acrylic adhesive
  • Microscope Slide-Wells are inert and stable under most laboratory conditions

Twin adhesive well stickers (1cm diameter) firmly adhere to microscope slides.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty