Would you like to log in again?
Disposable Adhesive Slide Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70366-14
102096-888PK 52.37 USD InStock
102096-888
Disposable Adhesive Slide Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Disposable adhesive slide grids are easy-to adhere stickers having side-by-side twin grids for microscope slides.
  • Compatible with different styles of microscopes
  • Use with dry specimens or suspended specimens
  • Chemically Inert - made of polyester with an acrylic adhesive
  • Slide-Grids are inert and stable under most laboratory conditions
Count or measure cells and estimate the size or length of microscopic or semi-micro objects. Virtually all cells and specimens placed on top of slide-grids can be counted and/or sized. Separate cover slips can be placed over each slide-grid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty