Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70450-85 70428 70441-20 70440-100 70442 70426 70444 70449 70424 70443 70448 70443-100 70441 70444-20 70427 70442-75 70429 70448-75 70425 70440 70449-85 70450
102091-140PK 119.72 USD InStock
102091-140 102091-150 102091-142 102091-152 102091-148 100499-906 100501-090 102091-138 102091-144 102091-146 100499-904 102096-902 100489-816 100489-814 102096-908 100501-092 102096-896 102096-894 102096-912 102096-900 102096-910 102096-898
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
Useful in all areas of cell culture including scale-up and cloning.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Nunclon™ Delta certified
  • Multidish 6 is available coated with Poly-D-Lysine or Collagen
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty