Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Willco-dish Kit-Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70665-22 70665-04 70665-20 70665-08 70665-13 70665-11 70665-03 70665-10 70665-07 70655-22 70665-06 70665-01 70665-09 70665-16 70665-21 70665-05 70665-02 70665-15 70665-12 70665-14
102097-124EA 380.82 USD InStock
102097-124 102097-102 102097-112 102097-104 102097-126 102097-114 102097-128 102097-106 102097-116 102097-108 102097-118 102097-094 102097-096 102097-098 102097-120 102097-122 102097-100 102097-110 102097-090 102097-092
Willco-dish Kit-Dishes, Electron Microscopy Sciences
Culture Dishes Petri Dishes
The WillCo-dish® KIT dishes, of which there are four types in polystyrene (PS) and all four in both transparent as well as in black walled dishes, are designed to offer Scientists the opportunity, to use any chosen material as well as different thicknesses, for the bottom of a dish.
  • Dish (Polystyrene or polycarbonate surrounds and -lids, clean room molded, 500 units each)
  • Pressure sensitive Double Sided [Medical] Adhesive rings, FDA approved (500 units)
  • Glass coverslips: German glass, 0.17 mm. - # 1.5 coverslips (500 units), # 1.5 on request
  • One pair of tweezers, to protect fingers from these thin, razor-sharp glass coverslips
  • "Assembly Devices" are optional for a quick, accurate and safe positioning and assembly of the glass coverslip and the adhesive ring
The WillCo-dish® KIT Glass Bottom dishes are composed of several components, each of the highest quality of their kind. Below please find a summery and description of these components.
Assembly Device: The 2 part aluminum assembly device comes separate, because you need to purchase it only once per type, in a life time. Again, using an assembly device with the KIT-dishes makes assembling these fine dishes easy, accurate and safe, also saving a lot of time and funds. The glass bottom of these dishes is always flush with the warming-stage and flat, a requirement for high quality imaging. These features ensure even distribution of the temperature inside the dish, as well as great imaging results.
As a standard, German glass Borosilicate cover slips are offered in Thickness: # 1.0 (130 – 160 micron), as well as # 1.5 (160 – 190 micron).
Ordering information: All kits are packed with dish, glass, and adhesive rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty