Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Petri Dishes — Sterile Petri Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70678-20 70678-40 70678-60 70678-30 70678-10 70678-50 70678-70
102097-170CS 258.36 USD InStock
102097-170 102097-162 102097-172 102097-164 102097-174 102097-166 102097-168
Petri Dishes — Sterile Petri Dishes, Electron Microscopy Sciences
Culture Dishes Petri Dishes
Made of polystyrene. Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by “corona effect”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty