Would you like to log in again?
Gloves, Latex, Surgeons, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71114-01 71115-01 71115-20 71114-20 71116-01 71116-20
102097-390CS 235.85 USD InStock
102097-390 102097-406 102097-396 102097-388 102097-398 102097-404
Gloves, Latex, Surgeons, Electron Microscopy Sciences
Gloves
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection.
  • Economical
  • Ambidextrous style
  • Beaded cuff
Chemically resistant to acids, alcohols, ketones and alkalis. Non-sterile.
Available either with powdered (biologically cornstarch dusting, ~2 mg/glove) or powder-free gloves.
Packaging: Packed in a convenient dispenser box, 100/box.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty