Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS Grid Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71146-02 71146-01
102097-418EA 16.2 USD InStock
102097-418 102097-420
EMS Grid Box, Electron Microscopy Sciences
Boxes Storage Boxes
The EMS50 and EMS100 TEM grid storage boxes are used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage for standard grids that are 3.05 mm in diameter.
  • Dimensions for both boxes are: 75×55×6.5 mm
  • Anti static treated
The EMS 50 and EMS100 capacity inexpensive grid storage boxes. The box has a simple number/letter combination printed on the side of the body.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty