Would you like to log in again?
Grid Transporting Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71173-01
102097-450EA 121.62 USD InStock
102097-450
Grid Transporting Box, Electron Microscopy Sciences
Boxes Transport Boxes
This unique Box allows for the storage and transportation of grids without any worry of them moving around or being damaged.
The Plate is made from Silicone and has 54 individual compartments 10 mm in size. Each compartment can hold 3 grids and the grids can be easily picked up from the Silicone surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty