Would you like to log in again?
Grid Staining Matrix System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71179-01 71179-08 71179-07 71179-06 71179-05
102097-460EA 63.92 USD InStock
102097-460 102097-452 102097-454 102097-456 102097-458
Grid Staining Matrix System, Electron Microscopy Sciences
Staining Boxes
The Matrix system has a simply alpha-numeric identification system. The unit is not solvent or chemically resistant to acids so all stains should be aqueous based only.
  • Stains up to 25 grids at one time or as little as one
The system requires very little stain and you may use different vessels for each stain.
Delivery information: Each system includes the Matrix Body and 2 each of the Staining Vessels (red and blue).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty