Would you like to log in again?
Slide Warmer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71320-220 71319-220
102097-470EA 590.83 USD InStock
102097-470 102097-478
Slide Warmer, Electron Microscopy Sciences
Slide Warmers/Dryers
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes.
  • Thermostatic heat control
  • Anodized black surface provides contrast
  • LED temperature display
  • Thermostat setting range from room temperature to 70 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty