Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

StainTray - Slide Staining System, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

71398-B 71397-B 71398-C 71396-C 71397-C 71396-B 71399-B 71399-C
102097-506EA 397.72 USD InStock
102097-506 101413-418 101413-416 102097-500 101413-420 101413-414 102097-502 102097-504
StainTray - Slide Staining System, Electron Microscopy Sciences
Jars Staining Jars
An ideal device for immunohistochemistry slide staining. The StainTray™ has a black base, made from tough ABS plastic, withstanding a wide range of chemicals (except chlorinated hydrocarbons). It will accept up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold the slides even if the tray is held at an angle.

  • Suitable for routine staining requiring a humid chamber
  • Ideal for hematology, cytology and microscopy laboratories
  • 20-Slide unit with cover: 15 x 93/8 x 13/4" H (38 x 24 x 4.5 cm H)
  • 10-Slide unit with cover: 93/8 x 93/8 x 1" (24 x 24 x 4.5 cm H)

When humidity is needed, the wells between the rails will hold the water securely without splashing. Rails are raised not only to avoid water touching the slides but also make the slides more easily retrievable. The base will also hold excess stain solution that drips from the slides. Four rubber feet add to the base for stability. Units are stackable to save space.

A clear cover is available that allows for visual examination. It is made from PETG with temperature a range of -20 to 60 °C. A black cover is available for fluorescent work. It is made from ABS with a temperature range of -80 to 80 °C.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty