Would you like to log in again?
Digital Vortex-Genie 2, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71838-BR 71838-20 71838-EU 71838-SW 71838-AU 71838-10 71838-12
102097-714EA 883.95 USD InStock
102097-714 102097-716 102097-708 102097-718 102097-720 102097-710 102097-712
Digital Vortex-Genie 2, Electron Microscopy Sciences
Shakers and Mixers Vortex Mixers
Additional accessories for the Vortex Genie® 2 Digital available.
Features digital control and display. Uses shaking or vortexing action. Variable speed control allows slow speed shaking action up to high-speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. Multi-Task Versatility. A broad range of attachments of various shapes, sizes, and materials allows for mixing of almost all common tubes or containers for Hands-Free or Touch operation. Digital speed control and display. Provides accuracy, reproducibility and repeatability. Timer Function. Touch on for 1-99 seconds. Hands-Free for 1-99 minutes or continuous. Does not walk. Tough metal housing combined with "just enough weight", provides a stable platform for all types of mixing. Unmatched Reliability. Many years of proven performance in laboratories all over the world.
Additional accessories for the Vortex Genie® 2 Digital available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty