Would you like to log in again?
Saw Blades and Frames, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72009 72015-01 72009-10 72015-10 72015-20
102091-240EA 39.34 USD InStock
102091-240 102097-790 102097-784 102097-786 102097-788
Saw Blades and Frames, Electron Microscopy Sciences
Razors and Blades
Swiss made saw blade used for cutting rubber, plastics, wax, plaster and similar materials which cuts in any direction.
SuperLok™ Diamond Wire starts with tensile core wire
A Swiss made saw frames are lightweight and perfectly balanced. Its ergonomic wooden handle reduces fatigue and provides greater dexterity. Patented blade-lock design eases blade insertion and maintains perfect alignment which eliminates wobble and reduces blade loss during use. It accepts saw blades up to 1.2 mm Ø and 130 mm long. This Frame is used for the Diamond Wire and Spiral Saw Blades.
This wire is specially drawn to Diamond Wire Technology specifications. It is heat-treated, stretched, and has a tensile strength in excess of 400,000 pounds. The diamond wire has an added electro copper sheath in which the diamonds, from 20 to 120 microns in size are impregnated. An additional overstrike is applied to provide greater bonding to the core. Diamond wire technology uses electronic measuring equipment to guarantee consistency in diamond impregnation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty