Would you like to log in again?
Scalpel Blades, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72049-21 72049-24 72049-11 72049-23 72049-10 72049-20 72049-15 72049-22 72049-25 72049-12
102097-836PK 30.72 USD InStock
102097-836 102097-846 102097-838 102097-828 102097-830 102097-840 102097-832 102097-842 102097-834 102097-844
Scalpel Blades, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpel Blades
Premiere® Blades are made from carbon steel, individually foil wrapped and gamma radiation sterilized.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty