Would you like to log in again?
Disposable Scalpel Sets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72052-22 72052-10 72052-11 72052-20 72052-15
102097-858PK 13.53 USD InStock
102097-858 102097-860 102097-852 102097-854 102097-856
Disposable Scalpel Sets, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Packaging: Packed 10 each /box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty