Would you like to log in again?
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72127-B12 72127-B1 72127-AS
101413-506EA 15.29 USD InStock
101413-506 101413-504 102097-872
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Pens Ink Pens
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins
  • With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it’s left uncapped
  • Once dry, the solvent base ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty