Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SuperNuova™ Digital Hot Plates, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SuperNuova™
72345-12 72345-20 72342-20 72342-52 72342-60 72342-12
102097-938EA 2209.35 USD InStock
102097-938 102097-916 102097-918 102097-940 102097-920 102097-922
SuperNuova™ Digital Hot Plates, Electron Microscopy Sciences
Heaters Hot Plates
Hot plates with rugged low profile cast aluminum body and ceramic or aluminum top plate.

  • Warning system deters accidental contact with hot surface
  • Remote probe allow for accurate reading of solution temperature to 0.1 °C for temperature below 100 °C
  • User-adjustable over-temperature protection prevents runaway temperature conditions
  • Rugged low profile cast aluminum body provides stability and durability
  • Unit designed provides protection of internal components in case of spillage
  • Available in 7 x 7" and 10 x 10" sizes

Features:
- Accommodates sample weights up to 35 lbs (19.5kg) on the 10 x 10" models, 25 lbs (11.3kg) on 7 x 7" models
- Integral ring stand holder accommodates 0.5" (1.3cm) diameter support rod
- Unique, single knob design controls heating. Set points go up slowly when the knob is rotated slowly, faster as the knob is rotated faster. The knob then is locked out to avoid accidental changes
- RS232 port enables you to output time count, temperature set points, top surface/remote probe temperature, to a printer or computer
- All models supplied with 6" type K probe, detachable cord and retention bracket that prevents cord from accidentally removed.
- Easy-to-read digital displays indicate temperature settings adjustable in 1 °C increments
- Designed to provide superior control of temperatures below 100 °C
- User programmable preset buttons allow for up to four often-used temperatures to be recalled at the touch of a single button. Ideal for lab that perform repetitive procedures
- Hot surface alert systems protect from accidental burns
- Walk away timer shuts off heating after a preset user-defined interval
- Independently adjustable over temperature protection circuit limits top plate temperature and prevent run away conditions
- Calibration mode allows remote probe to be calibrated to external standard and at any temperature the user selects

Ideal for cold room application. Can be used in a 0 to 27°C, 80% relative humidity, non-condensation environment

Choose from Ceramic or Aluminum Top:
- Rugged aluminum surface provides uniform heat distribution and added durability to sustain point impacts that might break ceramic or glass surface.
- Seamless reflective white ceramic top enhances sample visibility and provides an easy-to-clean and alkali/acid resistant surface.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty