Would you like to log in again?
MegaMag Genie™ Magnetic Stirrers, 230 V
Supplier: Electron Microscopy Sciences
72400-BR 72401-EU 72401-AU 72400-AU 72400-20 72400-EU 72401-20 72401-SW 72401-BR 72400-SW
102098-008EA 1504.48 USD InStock
102098-008 102098-006 102098-004 102097-990 102098-012 102098-010 102097-992 102097-994 102097-996 102097-998
MegaMag Genie™ Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
The powerful single-position MegaMag Genie and Digital MegaMag Genie Large volume magnetic stirrer that provids the optimal stirring for large volume containers.
  • 230 V model
Both analog and digital feedback (true speed) versions are available.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (102097-988 or 102098-002) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty