Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
MegaMag Genie®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72400-12 72401-12
102098-002EA 1504.48 USD InStock
102098-002 102097-988
MegaMag Genie®, Electron Microscopy Sciences
Stirrers Magnetic Stirrers
The powerful single-position MegaMag Genie and Digital MegaMag Genie Large volume magnetic stirrer that provids the optimal stirring for large volume containers.
Both analog and digital feedback (true speed) versions are available.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty