Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MultiMagStir Genie Magnetic Stirrers, 230 V
Supplier: Electron Microscopy Sciences
72405-EU 72405-20 72405-BR 72405-AU
102098-046EA 1711.92 USD InStock
102098-046 102098-044 102098-042 102098-040
MultiMagStir Genie Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
  • 230 V model
  • The revolutionary four position MultiMagStir Genie provides low to high speed stirring and editable memory profiles
  • Microprocessor controls offer precision and reproducibility without internal heat build-up
  • Customize your application with the four-position MultiMagStir Genie!
  • MultiMagStir Genie may be used with up to four 1 L flasks/beakers or three 1 L spinner flasks and a maximum sample weight of 10.0 kg
Speed range of 10 to 1200 rpm including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers. Three power levels are available for various sample viscosities. Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction. Manual mode provides stir, stir reverse, and time functions. Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to nine distinct steps. Up to four stirring programs can be saved. Automatic mode recalls and executes one of up to four user-defined protocols.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (102098-038) is also available, please see separate product page.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty