Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Hitachi S-450 SEM Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75600 75610-D 75620-D 76470 75610 75600-D 76471 75620
100488-810PK 157 USD InStock
100488-810 102098-168 100488-724 102098-166 100488-726 100501-854 102098-170 100488-808
Hitachi S-450 SEM Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
M4 Tapered hole, female thread.
Head 15 mm (5/8"), Pin height: 6 mm (1/4").
Ordering information: Comes plain or with 3 divisions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty