Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Slotted Head SEM Mount, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
75636-20 75636-40 75636-60 75636-90 75636-10 75636-50 75636-70 75636-80 75636-30
102098-228PK 3367.02 USD InStock
102098-228 102098-216 102098-226 102098-214 102098-224 102098-222 102098-220 102098-230 102098-218
Slotted Head SEM Mount, Electron Microscopy Sciences
Specimen Mounting Supplies
AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan Slotted Head

  • Aluminum Mounts – Made from ultra-pure aluminum

Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan Slotted Head
Head: 18 mm. Pin diameter 3.2 mm (1/8"). Pin height: 8 mm.

AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan Slotted Head
Head Diameter: 50 mm (2"). Pin diameter 3.2 mm (1/8").

AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan, Aluminum, Slotted Head
Head Diameter: 63 mm (2.48"). Pin diameter 3.2 mm (1/8").

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty